Learn More
Grainger G5371 ProtCase 4 7/64 in PwrClwLtcSys/PdLk Ylw
Catalog No. 502767534
Encompass_Preferred
The Grainger 16Z326 is a Westward 3/8-inch drive deep impact socket set. This 12-piece set includes sizes ranging from 10mm to 19mm as well as a 3-inch extension. The sockets are made from chrome vanadium steel for durability and have a black phosphate finish to resist corrosion. The sockets feature a deep design for accessing fasteners in tight spaces. This socket set comes packaged in a molded case for convenient storage and organization. The Westward brand is owned by Grainger and focuses on quality tools at an affordable price point for professional users.
Catalog No. 50-276-7534 Supplier Grainger Supplier No. 16Z326
The Grainger 16Z326 is a Westward 3/8-inch drive deep impact socket set. This 12-piece set includes sizes ranging from 10mm to 19mm as well as a 3-inch extension. The sockets are made from chrome vanadium steel for durability and have a black phosphate finish to resist corrosion. The sockets feature a deep design for accessing fasteners in tight spaces. This socket set comes packaged in a molded case for convenient storage and organization. The Westward brand is owned by Grainger and focuses on quality tools at an affordable price point for professional users.