Learn More
Grainger G5371 ProtCase 4 7/64 in PwrClwLtcSys/PdLk Blk
Catalog No. 502766560
Encompass_Preferred
The Grainger 16Z324 is a 3M Scotch-Brite General Purpose Hand Pad that is 6 inches by 9 inches and features an open web construction. It has a medium duty abrasiveness level and is made of nylon and synthetic fibers with resin bonding. This hand pad's open web design allows dirt and particles to move through the pad during cleaning. It can be used for a variety of cleaning applications including prep work, paint removal, cleaning, and surface conditioning. The 6 x 9 inch size provides good coverage for cleaning jobs.
Catalog No. 50-276-6560 Supplier Grainger Supplier No. 16Z324
The Grainger 16Z324 is a 3M Scotch-Brite General Purpose Hand Pad that is 6 inches by 9 inches and features an open web construction. It has a medium duty abrasiveness level and is made of nylon and synthetic fibers with resin bonding. This hand pad's open web design allows dirt and particles to move through the pad during cleaning. It can be used for a variety of cleaning applications including prep work, paint removal, cleaning, and surface conditioning. The 6 x 9 inch size provides good coverage for cleaning jobs.