Learn More
Grainger G5374 ProtCase 4 7/64 in PwrClwLtcSys/PdLk Blk
Catalog No. 502767909
Encompass_Preferred
The Grainger 16Z425 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1/2" diameter shaft and 1-1/8" long. The motor is totally enclosed, fan cooled with a Class B insulation and operates on single phase 115V power. This electric motor can be used for various industrial, commercial and residential applications that require 1/2 HP at 1725 RPM. The motor dimensions are 6-7/16" diameter x 8-3/16" long and it weighs approximately 18 pounds.
Catalog No. 50-276-7909 Supplier Grainger Supplier No. 16Z425
The Grainger 16Z425 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1/2" diameter shaft and 1-1/8" long. The motor is totally enclosed, fan cooled with a Class B insulation and operates on single phase 115V power. This electric motor can be used for various industrial, commercial and residential applications that require 1/2 HP at 1725 RPM. The motor dimensions are 6-7/16" diameter x 8-3/16" long and it weighs approximately 18 pounds.