Learn More
Grainger G5379 ProtCase 4.80 in PwrClwLtcSys/PdLk Blk
Catalog No. 502767182
Encompass_Preferred
The Grainger ProtCase 4.80 in PwrClwLtcSys PdLk Blk is a 4.8 inch power claw latching system with a black powder coat finish. It is made from steel and has a push to close design with an integrated paddle handle and 3-point latching for secure closure. This industrial grade latch system can withstand up to 2500 lbs of static load when properly installed. It comes with mounting hardware and installation template for easy mounting on cabinets, doors or panels. Overall this heavy duty latch provides secure closure for industrial storage and equipment.
Catalog No. 50-276-7182 Supplier Grainger Supplier No. 16Z603
The Grainger ProtCase 4.80 in PwrClwLtcSys PdLk Blk is a 4.8 inch power claw latching system with a black powder coat finish. It is made from steel and has a push to close design with an integrated paddle handle and 3-point latching for secure closure. This industrial grade latch system can withstand up to 2500 lbs of static load when properly installed. It comes with mounting hardware and installation template for easy mounting on cabinets, doors or panels. Overall this heavy duty latch provides secure closure for industrial storage and equipment.