Learn More
Grainger G5379 ProtCase 4.80 in PwrClwLtcSys/PdLk Ylw
Catalog No. 502768130
Encompass_Preferred
The Grainger 16Z605 is a Westward brand 1/4-inch drive socket set. It contains both SAE and metric sockets ranging from 4mm to 15mm and 1/8 inch to 9/16 inch. The set has a total of 38 pieces and comes in a molded case for storage and portability. The sockets are made from chrome vanadium steel for durability and have a matte finish to resist corrosion. This socket set is ideal for automotive and machinery repair and maintenance.
Catalog No. 50-276-8130 Supplier Grainger Supplier No. 16Z605
The Grainger 16Z605 is a Westward brand 1/4-inch drive socket set. It contains both SAE and metric sockets ranging from 4mm to 15mm and 1/8 inch to 9/16 inch. The set has a total of 38 pieces and comes in a molded case for storage and portability. The sockets are made from chrome vanadium steel for durability and have a matte finish to resist corrosion. This socket set is ideal for automotive and machinery repair and maintenance.