Learn More
Grainger G5383 ProtCase 5 29/32 in PwrClwLcSys/PdLk Blk
Catalog No. 502767757
Encompass_Preferred
The Grainger Approved 16Z728 is a 1/2 HP, 115V, 1-phase, 3450 RPM, 56C frame, open drip proof, ball bearing, C-face electric motor. It has a NEMA standard 48Y frame with a 1.25" shaft diameter and 1.38" shaft height. The motor is totally enclosed, fan cooled (TEFC) with a cast iron body, aluminum end brackets, and pre-lubricated ball bearings. It can be used for general purpose applications and is UL listed. The motor has a full load current of 7.2 Amps.
Catalog No. 50-276-7757 Supplier Grainger Supplier No. 16Z728
The Grainger Approved 16Z728 is a 1/2 HP, 115V, 1-phase, 3450 RPM, 56C frame, open drip proof, ball bearing, C-face electric motor. It has a NEMA standard 48Y frame with a 1.25" shaft diameter and 1.38" shaft height. The motor is totally enclosed, fan cooled (TEFC) with a cast iron body, aluminum end brackets, and pre-lubricated ball bearings. It can be used for general purpose applications and is UL listed. The motor has a full load current of 7.2 Amps.