Learn More
Grainger G5385 ProtCase 6 7/64 in PwrClwLtcSys/PdLk Ylw
Catalog No. 502766500
Encompass_Preferred
The Grainger Approved 16Z807 is a 1/2 HP, 115V, 1725 RPM, single phase, open drip proof motor with a 48 frame size. It has a 1/2" diameter shaft with a 1/4" keyway and features sleeve bearings for quiet operation. This motor is thermally protected and suitable for continuous duty applications. With its compact size, it can be used for powering pumps, fans, conveyors, and other industrial equipment. The motor is CSA certified and backed by a 1 year warranty from Grainger.
Catalog No. 50-276-6500 Supplier Grainger Supplier No. 16Z807
The Grainger Approved 16Z807 is a 1/2 HP, 115V, 1725 RPM, single phase, open drip proof motor with a 48 frame size. It has a 1/2" diameter shaft with a 1/4" keyway and features sleeve bearings for quiet operation. This motor is thermally protected and suitable for continuous duty applications. With its compact size, it can be used for powering pumps, fans, conveyors, and other industrial equipment. The motor is CSA certified and backed by a 1 year warranty from Grainger.