Learn More
Grainger Sargent Surface Vertical Rod Exit Device, Stainless Steel, Grade 1, 1 ea.
Catalog No. 502785958
Encompass_Preferred
The 8700 Series surface vertical rod exit device is designed for demanding single and double door applications requiring robust security. This Grade 1 ANSI/BHMA rated device provides reliable two-point latching with top and bottom bolts, featuring adjustable latching through the center case. It is compatible with wood or metal doors and includes options for hurricane-resistant or tornado-resistant configurations.
  • ANSI/BHMA A156.3 Grade 1 certified for strength and cycle durability.
  • UL10C fire and UL305 panic listed.
  • Suitable for 44 mm (1.75 in.) minimum door thickness.
  • Projection of 76 mm (3 in.) in neutral position and 54 mm (2.125 in.) when depressed.
  • Constructed with 13 mm (0.5 in.) rods in brass, bronze, or stainless steel.
  • Features hex key dogging on non-fire rated devices.
  • Accommodates door widths from 61 cm (24 in.) to 122 cm (48 in.) depending on rail size.
Catalog No. 50-278-5958 Supplier Grainger Supplier No. 18F593
The 8700 Series surface vertical rod exit device is designed for demanding single and double door applications requiring robust security. This Grade 1 ANSI/BHMA rated device provides reliable two-point latching with top and bottom bolts, featuring adjustable latching through the center case. It is compatible with wood or metal doors and includes options for hurricane-resistant or tornado-resistant configurations.
  • ANSI/BHMA A156.3 Grade 1 certified for strength and cycle durability.
  • UL10C fire and UL305 panic listed.
  • Suitable for 44 mm (1.75 in.) minimum door thickness.
  • Projection of 76 mm (3 in.) in neutral position and 54 mm (2.125 in.) when depressed.
  • Constructed with 13 mm (0.5 in.) rods in brass, bronze, or stainless steel.
  • Features hex key dogging on non-fire rated devices.
  • Accommodates door widths from 61 cm (24 in.) to 122 cm (48 in.) depending on rail size.