Learn More
Grainger SARGENT Surface Vertical Rod Exit Device, Stainless Steel, 36 in., 1 ea.
Catalog No. 502786395
Encompass_Preferred
The 8700 Series Surface Vertical Rod Exit Device is a robust, Grade 1 security solution engineered for commercial and institutional environments. Designed for single and double door applications, this wide stile touchbar device ensures reliable, two-point latching through adjustable top and bottom rods. It exceeds ANSI/BHMA requirements for cycle and strength, providing dependable performance in high-traffic settings.
  • Two-point latching system with top and bottom adjustability via center case
  • Constructed with durable stainless steel in a satin finish
  • Includes standard wood and machine mounting screws
  • Hex key dogging provided as standard on non-fire-rated devices
  • Compatible with wood or metal doors with 1-3/4 in. thickness
  • Minimum stile width: 4-1/2 in. with trim; 3-1/2 in. without trim
  • Designed for door widths from 33 in. to 36 in.
Catalog No. 50-278-6395 Supplier Grainger Supplier No. 18F596
The 8700 Series Surface Vertical Rod Exit Device is a robust, Grade 1 security solution engineered for commercial and institutional environments. Designed for single and double door applications, this wide stile touchbar device ensures reliable, two-point latching through adjustable top and bottom rods. It exceeds ANSI/BHMA requirements for cycle and strength, providing dependable performance in high-traffic settings.
  • Two-point latching system with top and bottom adjustability via center case
  • Constructed with durable stainless steel in a satin finish
  • Includes standard wood and machine mounting screws
  • Hex key dogging provided as standard on non-fire-rated devices
  • Compatible with wood or metal doors with 1-3/4 in. thickness
  • Minimum stile width: 4-1/2 in. with trim; 3-1/2 in. without trim
  • Designed for door widths from 33 in. to 36 in.