Learn More
Grainger Sargent Surface Vertical Rod Exit Device, Grade 1, Dark Oxidized Satin Bronze, 33 in. to 36 in. Width, 1 ea
Catalog No. 502786397
Encompass_Preferred
The 80 Series Surface Vertical Rod Exit Device is a high-performance, Grade 1 mechanical exit device designed for durability and security in wood or metal door applications. This device features a minimalist pushpad design, vandal-resistant construction, and two-point latching with top and bottom adjustability, making it suitable for commercial, medical, and educational environments.
  • ANSI/BHMA A156.3 Grade 1 certified for cycle and strength
  • UL10C fire and UL305 panic listed
  • Hex key dogging for standard non-fire rated devices
  • Surface vertical rod configuration with 3/4 in. stainless steel latchbolt throw
  • Designed for 33 in. to 36 in. door widths with a 4-1/2 in. minimum stile width
  • Pushbar projection of 3 in. (76 mm) neutral to 2-1/8 in. (54 mm) depressed
  • Supplied standard with wood and machine screws for installation
Catalog No. 50-278-6397 Supplier Grainger Supplier No. 18F598
The 80 Series Surface Vertical Rod Exit Device is a high-performance, Grade 1 mechanical exit device designed for durability and security in wood or metal door applications. This device features a minimalist pushpad design, vandal-resistant construction, and two-point latching with top and bottom adjustability, making it suitable for commercial, medical, and educational environments.
  • ANSI/BHMA A156.3 Grade 1 certified for cycle and strength
  • UL10C fire and UL305 panic listed
  • Hex key dogging for standard non-fire rated devices
  • Surface vertical rod configuration with 3/4 in. stainless steel latchbolt throw
  • Designed for 33 in. to 36 in. door widths with a 4-1/2 in. minimum stile width
  • Pushbar projection of 3 in. (76 mm) neutral to 2-1/8 in. (54 mm) depressed
  • Supplied standard with wood and machine screws for installation