Learn More
Cayman Chemical G6P Stndard 150UG
SDP
Catalog No. 502892478
Encompass_Preferred
An internal standard for the quantification of phosmet by GC- or LC-MS
Catalog No. 50-289-2478 Supplier Cayman Chemical Supplier No. 700752150 UG
An internal standard for the quantification of phosmet by GC- or LC-MS