Learn More
Aviva Systems Biology Corp G6pc Antibody - N-terminal region (ARP44223_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44223_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against G6pc. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AW107337, G6Pase, G6pt, Glc-6-PaseApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-131