Learn More
Epigentek Inc G6PC Polyclonal Antibody, HRP Conjulated (50 ul)
SDP
Supplier:  Epigentek Inc A62623050
Encompass_Preferred
HRP Polyclonal Antibody Raised in Rabbit. Immunogen: Recombinant Human Glucose-6-phosphatase protein (82-117AA). Tested Applications: ELISA. Reactivity: Human. Alternative Names: Glucose-6-phosphatase, G-6-Pase, G6Pase, Glucose-6-phosphatase alpha, G6Pase-alpha, G6PC, G6PT.
Catalog No. 50-123-9295