Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC G6PD activator AG1 | 421581-52-4 | 98.2% | 438.65 | C24H30N4S2 | 10MG
SDP

Catalog No. 5000237065
Encompass_Preferred

G6PD activator AG1 is a small-molecule research compound that selectively activates glucose-6-phosphate dehydrogenase (G6PD). It increases G6PD activity, elevates cellular glutathione, reduces reactive oxygen species, and has demonstrated protection against drug-induced hemolysis in cellular and erythrocyte assays.

  • Activates glucose-6-phosphate dehydrogenase with EC50 ≈ 3 μM.
  • Increases GSH and reduces ROS in cell-based assays.
  • Reduces hemolysis in human erythrocyte models.
  • Suitable for biochemical and cell-based studies of oxidative stress.
  • Molecular formula C24H30N4S2, molecular weight 438.65.
  • High purity; verify batch certificate (HPLC/LC-MS) for exact value.
  • Store at 4°C and protect from light; prepare solutions fresh due to limited stability.

Catalog No. 50-002-37065 Supplier Medchemexpress LLC Supplier No. HY12396210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

G6PD activator AG1 is a small-molecule research compound that selectively activates glucose-6-phosphate dehydrogenase (G6PD). It increases G6PD activity, elevates cellular glutathione, reduces reactive oxygen species, and has demonstrated protection against drug-induced hemolysis in cellular and erythrocyte assays.

  • Activates glucose-6-phosphate dehydrogenase with EC50 ≈ 3 μM.
  • Increases GSH and reduces ROS in cell-based assays.
  • Reduces hemolysis in human erythrocyte models.
  • Suitable for biochemical and cell-based studies of oxidative stress.
  • Molecular formula C24H30N4S2, molecular weight 438.65.
  • High purity; verify batch certificate (HPLC/LC-MS) for exact value.
  • Store at 4°C and protect from light; prepare solutions fresh due to limited stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.