Learn More
Medchemexpress LLC G6PD activator AG1 | 421581-52-4 | 98.2% | 438.65 g/mol | C24H30N4S2 | 1MG
SDP
Catalog No. 5000236061
Encompass_Preferred
G6PD activator AG1 is a small-molecule research reagent that selectively activates glucose-6-phosphate dehydrogenase (G6PD), reported to have an EC50 of approximately 3 μM. It is used in biochemical and cell-based studies to increase G6PD activity and to investigate cellular redox biology.
Catalog No. 50-002-36061 Supplier Medchemexpress LLC Supplier No. HY1239621MG
G6PD activator AG1 is a small-molecule research reagent that selectively activates glucose-6-phosphate dehydrogenase (G6PD), reported to have an EC50 of approximately 3 μM. It is used in biochemical and cell-based studies to increase G6PD activity and to investigate cellular redox biology.