Learn More
Medchemexpress LLC G6PD activator AG1 | 421581-52-4 | 99.5% | 438.65 g/mol | C24H30N4S2 | 50MG
SDP
Catalog No. 5000236441
Encompass_Preferred
G6PD activator AG1 is a small-molecule activator of glucose-6-phosphate dehydrogenase used as a biochemical probe in enzymatic and cellular assays. It is reported to selectively enhance G6PD activity (EC50 ≈ 3 μM) and is supplied as a solid with documented chemical identifiers and physicochemical data (CAS 421581-52-4; C24H30N4S2; MW 438.65 g/mol).
  • Potent activator of G6PD (EC50 ≈ 3 μM).
  • High reported purity (99.5%).
  • Available in multiple solid quantities, including 50 mg.
  • Suitable for studies of redox biology and enzyme modulation.
  • Provided with a downloadable product data sheet containing identifiers and properties.
Catalog No. 50-002-36441 Supplier Medchemexpress LLC Supplier No. HY12396250MG
G6PD activator AG1 is a small-molecule activator of glucose-6-phosphate dehydrogenase used as a biochemical probe in enzymatic and cellular assays. It is reported to selectively enhance G6PD activity (EC50 ≈ 3 μM) and is supplied as a solid with documented chemical identifiers and physicochemical data (CAS 421581-52-4; C24H30N4S2; MW 438.65 g/mol).
  • Potent activator of G6PD (EC50 ≈ 3 μM).
  • High reported purity (99.5%).
  • Available in multiple solid quantities, including 50 mg.
  • Suitable for studies of redox biology and enzyme modulation.
  • Provided with a downloadable product data sheet containing identifiers and properties.