Learn More
Medchemexpress LLC G6PD activator AG1 | 421581-52-4 | 99.5% | 438.65 g/mol | C24H30N4S2 | 5MG
SDP
Supplier:  Medchemexpress LLC HY1239625MG
Encompass_Preferred
G6PD activator AG1 is a small-molecule activator of glucose-6-phosphate dehydrogenase (G6PD) used in biochemical and cellular research. It increases G6PD activity and NADPH production, and has been applied in cell-based assays to reduce oxidative stress and erythrocyte hemolysis (reported EC50 ≈ 3 μM).
  • Activates glucose-6-phosphate dehydrogenase with an EC50 of approximately 3 μM.
  • Enhances NADPH production and reduces reactive oxygen species in cellular models.
  • Reduces erythrocyte hemolysis in relevant in vitro studies.
  • Provided as a solid research compound in multiple size options for assay flexibility.
  • High purity suitable for biochemical and cellular experiments (batch dependent).
Catalog No. 50-002-36229