Learn More
Cayman Chemical G6PD Activator AG1, Purity: ≥95%, CAS: 421581-52-4, 1 mg
SDP
Catalog No. 502919273
Encompass_Preferred
An activator of G6PDH (EC50 = 3.4 µM for G6PDHR459L); selectively induces the dimerization of G6PDHR459L over 6PGD, GAPDH, ALDH2, and ALDH3A1; decreases ROS levels in, and increases the viability of, primary human lymphocytes isolated from a patient carrying the G6PDHR459L mutation; inhibits chloroquine- or diamide-induced hemolysis in isolated human erythrocytes at 5 µM; decreases chloroquine-induced pericardial edema in zebrafish embryos at 1 µM; decreases tumor volume and weight and increases survival in an NCI H226 lung mesothelioma mouse xenograft model at 40 mg/kg
Catalog No. 50-291-9273 Supplier Cayman Chemical Supplier No. 396751 MG
An activator of G6PDH (EC50 = 3.4 µM for G6PDHR459L); selectively induces the dimerization of G6PDHR459L over 6PGD, GAPDH, ALDH2, and ALDH3A1; decreases ROS levels in, and increases the viability of, primary human lymphocytes isolated from a patient carrying the G6PDHR459L mutation; inhibits chloroquine- or diamide-induced hemolysis in isolated human erythrocytes at 5 µM; decreases chloroquine-induced pericardial edema in zebrafish embryos at 1 µM; decreases tumor volume and weight and increases survival in an NCI H226 lung mesothelioma mouse xenograft model at 40 mg/kg