Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANALYTICAL CONTROL SYSTEMS INC G6PDH Control, Normal, 0.5 ml per vial, 6 vials/pack, Stabilized oxyhemoglobin, Kinetic, Visual, Fluorescent, 5 days refrigerated, 21 days frozen, 2 years unreconstituted
SDP

Catalog No. NC1263511
Encompass

The ANALYTICAL CONTROL SYSTEMS INC HC-108 is a G6PDH control formulated to support the detection of G6PDH deficiencies and manage associated drug sensitivities. It is based on stabilized oxyhemoglobin, ensuring reliable performance across various assay types. The control maintains stability for 5 days when refrigerated, 21 days when frozen, and up to two years when lyophilized.

  • Product type: G6PDH control
  • Control level: Normal
  • Unit size: 0.5 ml per vial
  • Package quantity: 6 vials per pack
  • Base material: Stabilized oxyhemoglobin
  • Compatible with: Kinetic, Visual, Fluorescent assays
  • Stability (reconstituted, refrigerated): At least 5 days
  • Stability (reconstituted, frozen): 21 days
  • Stability (unreconstituted, lyophilized): Two years
  • Applications: Detection of G6PDH deficiencies, management of drug sensitivities

Catalog No. NC1263511 Supplier ANALYTICAL CONTROL SYSTEMS INC Supplier No. HC108
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The ANALYTICAL CONTROL SYSTEMS INC HC-108 is a G6PDH control formulated to support the detection of G6PDH deficiencies and manage associated drug sensitivities. It is based on stabilized oxyhemoglobin, ensuring reliable performance across various assay types. The control maintains stability for 5 days when refrigerated, 21 days when frozen, and up to two years when lyophilized.

  • Product type: G6PDH control
  • Control level: Normal
  • Unit size: 0.5 ml per vial
  • Package quantity: 6 vials per pack
  • Base material: Stabilized oxyhemoglobin
  • Compatible with: Kinetic, Visual, Fluorescent assays
  • Stability (reconstituted, refrigerated): At least 5 days
  • Stability (reconstituted, frozen): 21 days
  • Stability (unreconstituted, lyophilized): Two years
  • Applications: Detection of G6PDH deficiencies, management of drug sensitivities

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.