Learn More
Cayman Chemical G6PDH asy Reagent 100uL
SDP
Catalog No. 502893488
Encompass_Preferred
An internal standard for the quantification of ADB-5'Br-PINACA by GC- or LC-MS intended for research and forensic applications
Catalog No. 50-289-3488 Supplier Cayman Chemical Supplier No. 700753100 UL
An internal standard for the quantification of ADB-5'Br-PINACA by GC- or LC-MS intended for research and forensic applications