Learn More
Medchemexpress LLC HY-W107464 5mg , G6PDi-1 CAS:2457232-14-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW1074645MG
Encompass_Preferred
HY-W107464 5mg G6PDi-1 CAS: 2457232-14-1 G6PDi-1 is a reversible and non-competitive glucose-6-phosphate dehydrogenase (G6PD) inhibitor with an IC50 of 0.07 μM. G6PDi-1 depletes NADPH most strongly in lymphocytes. G6PDi-1 markedly decreases inflammatory cytokine production in T cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1229