Learn More
Selleck Chemical LLC G6PDi-1 S9617-5MG
SDP
Supplier:  Selleck Chemical LLC S96175MG
Encompass_Preferred
G6PDi-1 is an effective inhibitor of glucose-6-phosphate dehydrogenase (G6PD) G6PDi-1 depletes NADPH and decreases inflammatory cytokine production
Catalog No. 50-217-3478