Learn More
Medchemexpress LLC G7-18NATE TFA | 98.1% | 1417.50 | 1 MG
SDP
Catalog No. 5000423450
Encompass_Preferred
G7-18NATE TFA is a peptide inhibitor of Grb7. It binds to the Grb7-SH2 domain with micromolar affinity (Kd = 18.1 μM). G7-18NATE TFA inhibits cell proliferation, motility, cell invasion, and 3D culture formation in several cancer cell lines.
  • Peptide inhibitor of Grb7
  • Binds to the Grb7-SH2 domain with micromolar affinity (Kd = 18.1 μM)
  • Inhibits cell proliferation, motility, cell invasion, and 3D culture formation in several cancer cell lines
Catalog No. 50-004-23450 Supplier Medchemexpress LLC Supplier No. HYP10224A1MG
G7-18NATE TFA is a peptide inhibitor of Grb7. It binds to the Grb7-SH2 domain with micromolar affinity (Kd = 18.1 μM). G7-18NATE TFA inhibits cell proliferation, motility, cell invasion, and 3D culture formation in several cancer cell lines.
  • Peptide inhibitor of Grb7
  • Binds to the Grb7-SH2 domain with micromolar affinity (Kd = 18.1 μM)
  • Inhibits cell proliferation, motility, cell invasion, and 3D culture formation in several cancer cell lines