Learn More
Grainger LISTA Open Front Shelf Base Cabinet, Steel, Gray, 2 Shelves
Catalog No. 502769558
Encompass_Preferred
This open front shelf base cabinet provides durable, high-density storage for organized workspace management. Constructed from steel with a powder-coated finish, the unit is designed for stationary indoor use and features a sturdy frame with an open-face design for easy access to stored items.
  • Features 2 adjustable shelves with 440 lb. load capacity per shelf.
  • Shelf adjustment increments of 1 in.
  • Constructed from 18 ga. steel.
  • Assembled for immediate installation.
  • Overall dimensions of 56 1/2 in. W x 33 1/2 in. H x 28 1/2 in. D.
Catalog No. 50-276-9558 Supplier Grainger Supplier No. 13P581
This open front shelf base cabinet provides durable, high-density storage for organized workspace management. Constructed from steel with a powder-coated finish, the unit is designed for stationary indoor use and features a sturdy frame with an open-face design for easy access to stored items.
  • Features 2 adjustable shelves with 440 lb. load capacity per shelf.
  • Shelf adjustment increments of 1 in.
  • Constructed from 18 ga. steel.
  • Assembled for immediate installation.
  • Overall dimensions of 56 1/2 in. W x 33 1/2 in. H x 28 1/2 in. D.