Learn More
Eurofins Discoverx G9a, 50 µg. (EPI022)
Supplier:  Eurofins Discoverx EPI022
Encompass
G9a is purified, untagged recombinant G9a enzyme. This enzyme contains amino acids 913-1193. It retains its binding/enzymatic activity and is intended for use in epigenetic assays. This enzyme is provided as 1 mg/mL frozen in 50% glycerol. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-200-5199