Learn More
Cell Signaling Technology G9a/EHMT2 Monoclonal Antibody, Rabbit, 100 μl
SDP
Catalog No. 502732051
Encompass
The G9a/EHMT2 (D5R4R) Rabbit Monoclonal Antibody is a recombinant, monoclonal antibody generated by immunizing animals with a recombinant protein specific to the amino terminus of human G9a. This antibody recognizes endogenous levels of total G9a protein and is suitable for various applications, including chromatin immunoprecipitation and CUT&RUN assays.
  • Validated for Western blotting, immunoprecipitation, immunofluorescence, chromatin immunoprecipitation, and CUT&RUN assay
  • Reactivity with human, mouse, rat, and monkey samples
  • Recognizes endogenous G9a protein levels
  • Supplied in a formulation containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide
Catalog No. 50-273-2051 Supplier Cell Signaling Technology Supplier No. 68851S
The G9a/EHMT2 (D5R4R) Rabbit Monoclonal Antibody is a recombinant, monoclonal antibody generated by immunizing animals with a recombinant protein specific to the amino terminus of human G9a. This antibody recognizes endogenous levels of total G9a protein and is suitable for various applications, including chromatin immunoprecipitation and CUT&RUN assays.
  • Validated for Western blotting, immunoprecipitation, immunofluorescence, chromatin immunoprecipitation, and CUT&RUN assay
  • Reactivity with human, mouse, rat, and monkey samples
  • Recognizes endogenous G9a protein levels
  • Supplied in a formulation containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide