Learn More
Cayman Chemical G9alike protein human recomb
SDP
Catalog No. 502886464
Encompass_Preferred
Active recombinant N-terminal GST-tagged protein expressed in E. coli • G9a-like protein (GLP) is a SET domain-containing methyltransferases that specifically mono- and di-methylate histone H3 at lysine 9 (H3K9).
Catalog No. 50-288-6464 Supplier Cayman Chemical Supplier No. 1075550 UG
Active recombinant N-terminal GST-tagged protein expressed in E. coli • G9a-like protein (GLP) is a SET domain-containing methyltransferases that specifically mono- and di-methylate histone H3 at lysine 9 (H3K9).