Learn More
Pyramex™ G9WMG Safety Goggle
Catalog No. 19155324 Shop All Pyramex Safety Products Products
Change view
Click to view available options
ProductTypeSpecs: Wire mesh goggle
Catalog No.
19-155-324 ProductTypeSpecs: Wire mesh goggle
1 options
This item is not returnable. View return policy
Catalog No. 19-155-324 Supplier Pyramex™ Supplier No. G9WMG
Add to Cart
Add to Cart
This item is not returnable. View return policy
Low profile wire mesh goggle
  • Single wire mesh lens with PVC frame
  • Meets ANSI Z87.1-2015 wire screen requirements
  • Applications: forestry, landscaping, and agriculture
  • For work environments with extreme heat and high humidity
  • Low profile design with a wide elastic strap provides a secure fit
  • Wire mesh allows proper airflow and protects against flying debris
  • Soft rubber gasket seals out unwanted particles and is comfortable for all day wear
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Product Type Wire mesh goggle
    Frame Color Black
    Includes Elastic strap
    Lens Material Steel
    Frame Material PVC