Learn More
Medchemexpress LLC GA-017 | 2351906-74-4 | MFCD34676460 | 99.0% | C18H21N3O4 | 10MG
SDP
Catalog No. 5000258709
Encompass_Preferred
GA-017 is a potent, selective small-molecule inhibitor of LATS1 and LATS2 kinases that activates YAP/TAZ signaling, promotes cell proliferation in 3D culture, and enhances intestinal organoid formation. It is supplied as a high-purity research reagent for studies of cell signaling and stem cell/Wnt pathways.
  • Potent LATS1/2 inhibition with low-nanomolar IC50 values.
  • Promotes YAP/TAZ activation and nuclear translocation.
  • Enhances cell proliferation in 3D culture and organoid formation.
  • High purity suitable for biochemical and cellular assays.
  • Soluble in DMSO at approximately 20.8 mg/mL; may require warming and sonication.
  • Stable as a powder when stored at low temperatures for long-term use.
Catalog No. 50-002-58709 Supplier Medchemexpress LLC Supplier No. HY14708210MG
GA-017 is a potent, selective small-molecule inhibitor of LATS1 and LATS2 kinases that activates YAP/TAZ signaling, promotes cell proliferation in 3D culture, and enhances intestinal organoid formation. It is supplied as a high-purity research reagent for studies of cell signaling and stem cell/Wnt pathways.
  • Potent LATS1/2 inhibition with low-nanomolar IC50 values.
  • Promotes YAP/TAZ activation and nuclear translocation.
  • Enhances cell proliferation in 3D culture and organoid formation.
  • High purity suitable for biochemical and cellular assays.
  • Soluble in DMSO at approximately 20.8 mg/mL; may require warming and sonication.
  • Stable as a powder when stored at low temperatures for long-term use.