Learn More
Medchemexpress LLC HY-147082 5mg , GA-017 CAS:2351906-74-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1470825MG
Encompass_Preferred
HY-147082 5mg GA-017 CAS: 2351906-74-4 GA-017 is a potent and selective LATS1 and LATS2 (large tumor suppressor kinase 1/2) inhibitor, with IC50 values of 4.10 and 3.92 nM, respectively. GA-017 is an activator of cell proliferation. GA-017 promotes YAP/TAZ activation and nuclear translocation. GA-017 promotes cell growth under 3D culture conditions. GA-017 enhances the ex vivo formation of mouse intestinal organoids. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1874