Learn More
GA International GA International Direct Thermal Cryo Label, 25.4 x 54 mm, -196°C to +70°C, CryoSTUCK
Catalog No. NC3976812
Encompass_Preferred
CryoSTUCK Direct Thermal Cryo Labels rapidly label frozen vials and tubes without thawing. These durable labels resist harsh lab chemicals, water baths, and condensation across a wide temperature range.
  • Can be affixed to already frozen vials and containers as low as -80°C.
  • Permanent adhesive for cryogenic environments including liquid nitrogen and ultra-low freezers (-196°C to +70°C).
  • Withstands thawing in heated water baths.
  • Resistant to common lab chemicals like detergents, ethanol, and disinfectants.
  • Compatible with most direct thermal printers; no ribbons or ink required.
  • No cure time needed; immediate storage of vials after application.
Catalog No. NC3976812 Supplier GA International Supplier No. DTU284NOTC10.5WH
CryoSTUCK Direct Thermal Cryo Labels rapidly label frozen vials and tubes without thawing. These durable labels resist harsh lab chemicals, water baths, and condensation across a wide temperature range.
  • Can be affixed to already frozen vials and containers as low as -80°C.
  • Permanent adhesive for cryogenic environments including liquid nitrogen and ultra-low freezers (-196°C to +70°C).
  • Withstands thawing in heated water baths.
  • Resistant to common lab chemicals like detergents, ethanol, and disinfectants.
  • Compatible with most direct thermal printers; no ribbons or ink required.
  • No cure time needed; immediate storage of vials after application.