Learn More
Lonza Walkersville Gentamicin sulfate-Amphotericin (GA-1000) , 5 mL
Catalog No. NC0703289
Encompass
GA-1000 vial contains 30 mg/ml Gentamicin and 15 µg/ml Amphotericin. We recommend use at a 1:1000 ratio (i.e. 0.2 ml into a 200 ml bottle for a final concentration of 30 µg/ml Gentamicin and 15 ng/ml Amphotericin. GA-1000 is included in all Clonetics Primary Cell Culture Media Kits. This product is not suitable for human or veterinary use.Content:1 Vial Containing 5 mL GA-1000. 30 mg/mL Gentamicin and 15 µg/mL Amphotericin
Catalog No. NC0703289 Supplier Lonza Walkersville Supplier No. CC4083
GA-1000 vial contains 30 mg/ml Gentamicin and 15 µg/ml Amphotericin. We recommend use at a 1:1000 ratio (i.e. 0.2 ml into a 200 ml bottle for a final concentration of 30 µg/ml Gentamicin and 15 ng/ml Amphotericin. GA-1000 is included in all Clonetics Primary Cell Culture Media Kits. This product is not suitable for human or veterinary use.Content:1 Vial Containing 5 mL GA-1000. 30 mg/mL Gentamicin and 15 µg/mL Amphotericin