Learn More
Vector Laboratories Human GAA / Alpha-Glucosidase, Acid (Sandwich ELISA) ELISA Kit - LS-F13304
SDP
Catalog No. NC2873558
Encompass
LS-F13304 is a 96-well enzyme-linked immunosorbent assay (ELISA) for the Quantitative detection of Human GAA / Alpha-Glucosidase, Acid in samples of Serum. It is based upon a Sandwich assay principle and can be used to detect levels of GAA / Alpha-Glucosidase, Acid as low as 0.98 nanograms per millilter.
Catalog No. NC2873558 Supplier Vector Laboratories Supplier No. LSF133041
LS-F13304 is a 96-well enzyme-linked immunosorbent assay (ELISA) for the Quantitative detection of Human GAA / Alpha-Glucosidase, Acid in samples of Serum. It is based upon a Sandwich assay principle and can be used to detect levels of GAA / Alpha-Glucosidase, Acid as low as 0.98 nanograms per millilter.