Learn More
Enzo Life Sciences gamma-Aminobutyric acid monoclonal antibody (5A9) (100µl)
SDP
Supplier:  Enzo Life Sciences BMLGA11600100
Encompass_Preferred
Clone: 5A9. Isotype: IgG1. Immunogen: GABA conjugated to bovine serum albumin using glutaraldehyde. Species reactivity: Species independent. Specificity: Recognizes GABA. Species independent. Applications: IHC, WB. Formulation: Liquid. In PBS with 0.01M sodium azide.
Catalog No. 50-201-0890