Learn More
Cayman Chemical GABAA RECPTR 2 SubunIt Pol
SDP
Catalog No. 502938292
Encompass_Preferred
A diacylglycerol contains stearic acid at the sn-1 position and lauric acid at the sn-2 position a substrate for the rat diacylglycerol lipase DDHD2
Catalog No. 50-293-8292 Supplier Cayman Chemical Supplier No. 29276100 UL
A diacylglycerol contains stearic acid at the sn-1 position and lauric acid at the sn-2 position a substrate for the rat diacylglycerol lipase DDHD2