Learn More
Eurofins Discoverx ChemiScreen GABAB Receptor Membrane Preparation, 1 vial. (HTS119M)
Catalog No. 502005919
Encompass
The ChemiScreen GABAB Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of GABBR1 - GABBR2 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.
Catalog No. 50-200-5919 Supplier Eurofins Discoverx Supplier No. HTS119M
The ChemiScreen GABAB Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of GABBR1 - GABBR2 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.