Learn More
Enzo Life Sciences Gabaculine (5mg). CAS: 59556-17-1
SDP
Supplier:  Enzo Life Sciences BMLEA1350005
Encompass_Preferred
A potent, irreversible inhibitor of GABA transaminase. Alternative name: (+/-)-Amino-2,3-dihydrobenzoic acid . HCl. Purity: ≥95% (TLC). Solubility: Soluble in water (25 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0534