Learn More
Medchemexpress LLC Gabapentin | 60142-96-3 | 865286 | 98.0% | C9H17NO2 | 1 ML
SDP
Catalog No. 5000433549
Encompass_Preferred
Gabapentin is a GABA analog that inhibits neuronal Ca2+ influx and reduces neurotransmitter release. It is used to relieve neuropathic pain and is provided here as a 10 mM solution in 1 mL of water.
  • Inhibits neuronal Ca2+ influx
  • Reduces neurotransmitter release
  • GABA analog
  • Used to relieve neuropathic pain
  • Provided as a 10 mM solution
  • Solution is in 1 mL of water
Catalog No. 50-004-33549 Supplier Medchemexpress LLC Supplier No. HYA005710MM/1ML
Gabapentin is a GABA analog that inhibits neuronal Ca2+ influx and reduces neurotransmitter release. It is used to relieve neuropathic pain and is provided here as a 10 mM solution in 1 mL of water.
  • Inhibits neuronal Ca2+ influx
  • Reduces neurotransmitter release
  • GABA analog
  • Used to relieve neuropathic pain
  • Provided as a 10 mM solution
  • Solution is in 1 mL of water