Learn More
Medchemexpress LLC Gabapentin | 60142-96-3 | 98.0% | 171.24 | 10 MG
SDP
Catalog No. 5000365510
Encompass_Preferred
Gabapentin is a potent, orally active P/Q type Ca2+ channel blocker. It inhibits neuronal Ca2+ influx and reduces neurotransmitter release. As a GABA analog, it can be used to relieve neuropathic pain.
  • Inhibits neuronal Ca2+ influx
  • Reduces neurotransmitter release
  • Alleviates neuropathic pain
  • Inhibits K+-induced [Ca2+]i increase in synaptosomes
  • Decreases K+-evoked release of endogenous aspartate and glutamate in neocortical slices
  • Reduces K+-evoked [3H]-noradrenaline release in neocortical slices
Catalog No. 50-003-65510 Supplier Medchemexpress LLC Supplier No. 10MGHYA0057
Gabapentin is a potent, orally active P/Q type Ca2+ channel blocker. It inhibits neuronal Ca2+ influx and reduces neurotransmitter release. As a GABA analog, it can be used to relieve neuropathic pain.
  • Inhibits neuronal Ca2+ influx
  • Reduces neurotransmitter release
  • Alleviates neuropathic pain
  • Inhibits K+-induced [Ca2+]i increase in synaptosomes
  • Decreases K+-evoked release of endogenous aspartate and glutamate in neocortical slices
  • Reduces K+-evoked [3H]-noradrenaline release in neocortical slices