Learn More
Cayman Chemical Gabapentin-d4, Purity: ≥95%, CAS: 1185039-20-6, 1 mg
SDP
Catalog No. 502968307
Encompass_Preferred
An internal standard for the quantification of gabapentin by GC- or LC-MS
Catalog No. 50-296-8307 Supplier Cayman Chemical Supplier No. 182541 MG
An internal standard for the quantification of gabapentin by GC- or LC-MS