Learn More
Cayman Chemical GabapentIn-d4 500ug
SDP
Catalog No. 502967661
Encompass_Preferred
An internal standard for the quantification of gabapentin by GC- or LC-MS
Catalog No. 50-296-7661 Supplier Cayman Chemical Supplier No. 18254500 UG
An internal standard for the quantification of gabapentin by GC- or LC-MS