Learn More
Medchemexpress LLC Gabapentin-d6 hydrochloride | 1432061-73-8 | >98% | C9H12D6ClNO2 | 1 MG
SDP
Catalog No. 5000432319
Encompass_Preferred
Gabapentin-d6 hydrochloride is a deuterium-labeled version of Gabapentin hydrochloride. It is intended for research use only and is not sold to patients. This compound can be utilized as a tracer or as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. Stable heavy isotopes, including hydrogen, carbon, and other elements, are incorporated into drug molecules primarily for quantitation during the drug development process. Deuteration has garnered attention for its potential impact on the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-32319 Supplier Medchemexpress LLC Supplier No. HYA0057AS1MG
Gabapentin-d6 hydrochloride is a deuterium-labeled version of Gabapentin hydrochloride. It is intended for research use only and is not sold to patients. This compound can be utilized as a tracer or as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. Stable heavy isotopes, including hydrogen, carbon, and other elements, are incorporated into drug molecules primarily for quantitation during the drug development process. Deuteration has garnered attention for its potential impact on the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.