Learn More
Apexbio Technology LLC Gabapentin HCl 60142-95-2 20mg
SDP
Catalog No. 5000088746
Encompass_Preferred
Gabapentin HCl (CAS 60142-95-2) is a structurally related analog of gamma-aminobutyric acid (GABA) though it does not bind directly to GABA receptors Its primary mechanism involves binding to the 2 subunit of voltage-gated calcium channels in neuronal tissues resulting in decreased neurotransmitter release Originally investigated for antiepileptic properties gabapentin has been widely utilized in research exploring neuropathic pain modulation and neuronal excitability This molecule serves as a tool compound for studying calcium channel modulation and the pathophysiology of neurological disorders
Catalog No. 50-000-88746 Supplier Apexbio Technology LLC Supplier No. B152920
Gabapentin HCl (CAS 60142-95-2) is a structurally related analog of gamma-aminobutyric acid (GABA) though it does not bind directly to GABA receptors Its primary mechanism involves binding to the 2 subunit of voltage-gated calcium channels in neuronal tissues resulting in decreased neurotransmitter release Originally investigated for antiepileptic properties gabapentin has been widely utilized in research exploring neuropathic pain modulation and neuronal excitability This molecule serves as a tool compound for studying calcium channel modulation and the pathophysiology of neurological disorders