Learn More
Medchemexpress LLC Gabapentin hydrochloride | 60142-95-2 | 98.0% | C9H18ClNO2 | 1 ML
SDP
Catalog No. 5000432310
Encompass_Preferred
Gabapentin hydrochloride is a potent, orally active P/Q type Ca2+ channel blocker. It inhibits neuronal Ca2+ influx and reduces neurotransmitter release. This GABA analog can be used to relieve neuropathic pain.
  • Potent, orally active P/Q type Ca2+ channel blocker
  • Inhibits neuronal Ca2+ influx
  • Reduces neurotransmitter release
  • GABA analog for neuropathic pain relief
  • Improves spatial and emotional cognitive performance (in mice)
  • Exhibits analgesic effect and reduces writhing (in mice)
Catalog No. 50-004-32310 Supplier Medchemexpress LLC Supplier No. HYA0057A10MM/1ML
Gabapentin hydrochloride is a potent, orally active P/Q type Ca2+ channel blocker. It inhibits neuronal Ca2+ influx and reduces neurotransmitter release. This GABA analog can be used to relieve neuropathic pain.
  • Potent, orally active P/Q type Ca2+ channel blocker
  • Inhibits neuronal Ca2+ influx
  • Reduces neurotransmitter release
  • GABA analog for neuropathic pain relief
  • Improves spatial and emotional cognitive performance (in mice)
  • Exhibits analgesic effect and reduces writhing (in mice)