Learn More
Medchemexpress LLC Gabapentin hydrochloride | 60142-95-2 | 98.0% | C9H18ClNO2 | 100 MG
SDP
Catalog No. 5000432077
Encompass_Preferred
Gabapentin hydrochloride is a potent, orally active P/Q type Ca2+ channel blocker. It inhibits neuronal Ca2+ influx and reduces neurotransmitter release. This GABA analog can be used to relieve neuropathic pain.
  • Potent, orally active P/Q type Ca2+ channel blocker
  • Inhibits neuronal Ca2+ influx
  • Reduces neurotransmitter release
  • GABA analog for neuropathic pain relief
  • Inhibits K+-induced Ca2+ increase in synaptosomes
  • Decreases K+-evoked release of aspartate and glutamate
  • Reduces K+-evoked noradrenaline release
  • Improves cognitive performance in animal models
  • Provides analgesic effects in animal models
Catalog No. 50-004-32077 Supplier Medchemexpress LLC Supplier No. HYA0057A100MG
Gabapentin hydrochloride is a potent, orally active P/Q type Ca2+ channel blocker. It inhibits neuronal Ca2+ influx and reduces neurotransmitter release. This GABA analog can be used to relieve neuropathic pain.
  • Potent, orally active P/Q type Ca2+ channel blocker
  • Inhibits neuronal Ca2+ influx
  • Reduces neurotransmitter release
  • GABA analog for neuropathic pain relief
  • Inhibits K+-induced Ca2+ increase in synaptosomes
  • Decreases K+-evoked release of aspartate and glutamate
  • Reduces K+-evoked noradrenaline release
  • Improves cognitive performance in animal models
  • Provides analgesic effects in animal models