Learn More
Medchemexpress LLC Gabapentin hydrochloride | 60142-95-2 | >98.0% | C9H18ClNO2 | 200 MG
SDP
Catalog No. 5000433470
Encompass_Preferred
Gabapentin hydrochloride is a potent, orally active P/Q type Ca2+ channel blocker. It functions by inhibiting neuronal Ca2+ influx and reducing neurotransmitter release. This compound, which is a GABA analog, can be used to relieve neuropathic pain.
  • It is a potent calcium channel blocker.
  • It is orally active.
  • Inhibits neuronal calcium influx.
  • Reduces neurotransmitter release.
  • It is a GABA analog.
  • Can be used to relieve neuropathic pain.
Catalog No. 50-004-33470 Supplier Medchemexpress LLC Supplier No. HYA0057A200MG
Gabapentin hydrochloride is a potent, orally active P/Q type Ca2+ channel blocker. It functions by inhibiting neuronal Ca2+ influx and reducing neurotransmitter release. This compound, which is a GABA analog, can be used to relieve neuropathic pain.
  • It is a potent calcium channel blocker.
  • It is orally active.
  • Inhibits neuronal calcium influx.
  • Reduces neurotransmitter release.
  • It is a GABA analog.
  • Can be used to relieve neuropathic pain.