Learn More
Medchemexpress LLC Gabapentin hydrochloride | 60142-95-2 | 98.0% | C9H18ClNO2 | 500 MG
SDP
Catalog No. 5000432078
Encompass_Preferred
Gabapentin hydrochloride is a potent, orally active P/Q type Ca2+ channel blocker. It inhibits neuronal Ca2+ influx and reduces neurotransmitter release. It is also a GABA analog that can be used to relieve neuropathic pain.
  • Potent and orally active P/Q type Ca2+ channel blocker
  • Inhibits neuronal Ca2+ influx
  • Reduces neurotransmitter release
  • GABA analog for relieving neuropathic pain
  • For research use only
Catalog No. 50-004-32078 Supplier Medchemexpress LLC Supplier No. HYA0057A500MG
Gabapentin hydrochloride is a potent, orally active P/Q type Ca2+ channel blocker. It inhibits neuronal Ca2+ influx and reduces neurotransmitter release. It is also a GABA analog that can be used to relieve neuropathic pain.
  • Potent and orally active P/Q type Ca2+ channel blocker
  • Inhibits neuronal Ca2+ influx
  • Reduces neurotransmitter release
  • GABA analog for relieving neuropathic pain
  • For research use only