Learn More
Aviva Systems Biology Corp GABARAP Antibody - N-terminal region (ARP51265_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51265_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GABARAP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GABARAP. Alternate names: FLJ25768, MGC120154, MGC120155, MM46, ATG8AApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-60530