Learn More
Aviva Systems Biology Corp Gabarapl2 Antibody - C-terminal region (ARP52326_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52326_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Gabarapl2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Gef2, MGC108686, Gabarapl2ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-706