Learn More
ABclonal Technology GABARAPL2 Rabbit pAb
SDP
Catalog No. 5000106180
Encompass_Preferred
Enables ubiquitin protein ligase binding activity. Involved in negative regulation of proteasomal protein catabolic process and protein localization to endoplasmic reticulum. Located in Golgi membrane and autophagosome membrane.
Catalog No. 50-001-06180 Supplier ABclonal Technology Supplier No. A14855500UL
Enables ubiquitin protein ligase binding activity. Involved in negative regulation of proteasomal protein catabolic process and protein localization to endoplasmic reticulum. Located in Golgi membrane and autophagosome membrane.